4-Amino-3-fluorbenzylalkohol

Lieferant: BIONET RESEARCH

Synonyme: (4-Amino-3-fluorophenyl)methanol

FA-0632.1
BIOTFA-0632.1EA 0 EUR
BIOTFA-0632.1
4-Amino-3-fluorbenzylalkohol
4-Amino-3-fluorbenzylalkohol
Formel: C₇H₈FNO
Molecular Weight: 141,1429 g/mol
MDL: MFCD03094241
CAS-Nummer: 146019-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR