Natriumperiodat
Lieferant: G Bioscienes
|
Gefahr
|
G-Biosciences' Sodium meta -Periodate is a mild oxidizing agent that converts carbohydrates to activated active aldehydes
G-Biosciences' Sodium meta-Periodate is also suitable for oxidizing glycoproteins.
It is used in coupling to amines with cyanborohydride reduction.
Formel:
NaIO₄ Siedepunkt: 300 °C (1013 hPa) Schmelzpunkt: 175 °C Dichte: 3,87 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003534 CAS-Nummer: 7790-28-5 EINECS: 232-197-6 UN: 1479 ADR: 5.1,II Merck Index: 13,08714 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...