Dimethylsulfoxid, anhydrous

Lieferant: G Bioscienes

Synonyme: Methylsulfinylmethan, DMSO

BKC-17
BKC-17EA 188 EUR
BKC-17
Dimethylsulfoxid, anhydrous
Dimethylsulfoxid

Anydrous DMSO [Dimethyl sulfoxide (CH3)2SO] is offered for the solubilization of the water insoluble cross-linkers and modification reagents.

Formel: (CH₃)₂SO
Molecular Weight: 78.14 g/mol
Siedepunkt: 189
Schmelzpunkt: 18 °C
Dichte: 1.101 g/cm³ (20 °C)
Flammpunkt: 87
Lagertemperatur: Raumtemperatur
MDL: MFCD00002089
CAS-Nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR