2,5-Dihydroxy-1,4-dioxane Gemisch von Stereoisomeren 95%

Lieferant: BLD PHARMATECH
Achtung

Synonyme: 1,4-Dioxane-2,5-diol

BD132241
BLDPBD132241EA 0 EUR
BLDPBD132241
2,5-Dihydroxy-1,4-dioxane Gemisch von Stereoisomeren 95%
2,5-Dihydroxy-1,4-dioxane

Storage condition: Inert atmosphere, 2-8°C

Formel: C₄H₈O₄
Molecular Weight: 120,11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00012133
CAS-Nummer: 23147-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR