Human recombinant IL-1beta E. coli
Lieferant: BIOLEGEND INC
579409
BLEG579409EA 144 EUR
BLEG579409
Human recombinant IL-1beta E. coli
Proteine und Peptide
ELISA Standard 4-Pack contains four vials of recombinant human IL-1b at >5 ng per vial. Recommended for ELISA application only. Standards are calibrated against a BioLegend Master Calibrator.