VWR Chemicals Mucap test®, Salmonella identification
Lieferant: BIOLIFE ITALIANA
191500
BLIF191500EA
0
EUR
BLIF191500
VWR Chemicals Mucap test®, Salmonella identification
Medien für die Mikrobiologie
Additive und Supplemente für die Mikrobiologie
The Mucap test® allows the presumptive detection of Salmonella spp. Wood lamp with emission at 366 nm is required. Positive colonies on the plate show blue fluorescence.
- Easy
- Rapid