Tricin

Lieferant: BIOMOL RESEARCH LABORATORIES

Synonyme: N-(Tris(hydroxymethyl)methyl)glycin

22870-1 22870-2 22870-3
BMOL22870-1EA 0 EUR
BMOL22870-1 BMOL22870-2 BMOL22870-3
Tricin
Tricin
Formel: (HOCH₂)₃CNHCH₂CO₂H
Molecular Weight: 179.17 g/mol
Schmelzpunkt: 187
Dichte: 1.523 g/cm³ (19.85 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004277
CAS-Nummer: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR