n-Decyl-β-D-maltosid
Lieferant: BIOMOL RESEARCH LABORATORIES
50165-1
50165-2
BMOL50165-1EA
0
EUR
BMOL50165-1
BMOL50165-2
n-Decyl-β-D-maltosid
n-Decyl-β-D-maltosid
Formel:
C₂₂H₄₂O₁₁ |
MDL:
MFCD00061624 CAS-Nummer: 82494-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...