Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)

Lieferant: BIOMOL RESEARCH LABORATORIES

50200-1 50200-2 50200-3
BMOL50200-1EA 0 EUR
BMOL50200-1 BMOL50200-2 BMOL50200-3
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz)
Formel: C₁₀H₁₄N₅Na₂O₁₃P₃
MDL: MFCD00080339
CAS-Nummer: 987-65-5
EINECS: 213-579-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR