3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propansulfonat
Lieferant: BIOMOL RESEARCH LABORATORIES
|
Achtung
|
Synonyme:
3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropansulfonat, CHAPSO
50792-1
50792-2
BMOL50792-1EA
0
EUR
BMOL50792-1
BMOL50792-2
3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propansulfonat
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Formel:
C₃₂H₅₈N₂O₈S Molecular Weight: 630.89 g/mol Schmelzpunkt: 184…186 °C Lagertemperatur: Kühlschrank |
CAS-Nummer:
82473-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...