Nonyl-β-D-glucopyranosid

Lieferant: BIOMOL RESEARCH LABORATORIES

Synonyme: Nonyl-β-D-glucosid, NG

50862-1 50862-2
BMOL50862-1EA 0 EUR
BMOL50862-1 BMOL50862-2
Nonyl-β-D-glucopyranosid
Nonyl-β-D-glucopyranosid
Formel: C₁₅H₃₀O₆
Molecular Weight: 306.4 g/mol
MDL: MFCD00063300
CAS-Nummer: 69984-73-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR