n-Undecyl-β-D-maltosid

Lieferant: BIOMOL RESEARCH LABORATORIES

52693-1 52693-2
BMOL52693-1EA 0 EUR
BMOL52693-1 BMOL52693-2
n-Undecyl-β-D-maltosid
n-Undecyl-β-D-maltosid
Formel: C₂₃H₄₄O₁₁
CAS-Nummer: 170552-39-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR