n-Octyl-β-D-maltosid

Lieferant: BIOMOL RESEARCH LABORATORIES

54560-1 54560-2
BMOL54560-1EA 0 EUR
BMOL54560-1 BMOL54560-2
n-Octyl-β-D-maltosid
n-Octyl-β-D-maltosid
Formel: C₂₀H₃₈O₁₁
CAS-Nummer: 82494-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR