TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether) 10% in Wasser
Lieferant: BIOMOL RESEARCH LABORATORIES
|
Gefahr
|
54561-1
54561-2
54561-3
BMOL54561-1EA
0
EUR
BMOL54561-1
BMOL54561-2
BMOL54561-3
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether) 10% in Wasser
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)
Siedepunkt:
∼100 °C (1013 hPa) Dichte: ∼1,01 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00132505 CAS-Nummer: 9002-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...