D(+)-Trehalose Dihydrat
Lieferant: BIOMOL RESEARCH LABORATORIES
54688-2
BMOL54688-2EA
0
EUR
BMOL54688-2
D(+)-Trehalose Dihydrat
D(+)-Trehalose Dihydrat
Formel:
C₁₂H₂₂O₁₁·2H₂O Schmelzpunkt: 97…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071594 CAS-Nummer: 6138-23-4 EINECS: 202-739-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...