Ectoin
Lieferant: BIOMOL RESEARCH LABORATORIES
Synonyme:
(S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
55459-1
55459-3
55459-7
BMOL55459-1EA
0
EUR
BMOL55459-1
BMOL55459-3
BMOL55459-7
Ectoin
Ectoin
Formel:
C₆H₁₀N₂O₂ Molecular Weight: 142.1558 g/mol |
MDL:
MFCD03419286 CAS-Nummer: 96702-03-3 EINECS: 431-910-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...