n-Dodecyl-β-D-maltosid
Lieferant: BIOMOL RESEARCH LABORATORIES
|
Achtung
|
Synonyme:
DDM
56366-1
56366-2
56366.25
BMOL56366-1EA
0
EUR
BMOL56366-1
BMOL56366-2
BMOL56366.25
n-Dodecyl-β-D-maltosid
n-Dodecyl-β-D-maltosid
Formel:
C₂₄H₄₆O₁₁ Molecular Weight: 510.6 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00043012 CAS-Nummer: 69227-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...