Rat IgA ELISA Kit
Lieferant: BIOMOL RESEARCH LABORATORIES
E111-102
BMOLE111-102EA 1180 EUR
BMOLE111-102
Rat IgA ELISA Kit
Tests ELISAs
This ELISA is for the detection of rat IgA in serum or plasma. Due to idiotypic and allotypic variations, this product may not accurately quantitate hybridomas, monoclonals or myelomas. This kit contains sufficient components to quantitate rat IgA in up to 40 samples, tested in duplicate.