Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid 98, ultrapure
Lieferant: BIOMOL RESEARCH LABORATORIES
Synonyme:
(7Z,10Z,13Z,16Z,19Z)-docosa-7,10,13,16,19-pentaenoic acid, Clupanodonic acid
FA012-0010
BMOLFA012-0010EA
0
EUR
BMOLFA012-0010
Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid 98, ultrapure
Docosa-7Z,10Z,13Z,16Z,19Z-pentaenoic acid
Polyunsaturated acid
Formel:
C₂₂H₃₄O₂ Molecular Weight: 330.512 g/mol |
CAS-Nummer:
24880-45-3 |
Spezifikation Testergebnisse
Molecular Formula | C22H34O2 |
Molecular Weight | 330.5 g/mol |
Source | Synthetic |
CAS | 24880-45-3 |
Purity | ≥ 98% |
Appearance | Clear to pale yellow oil |
Solubility | Soluble in ethanol, hexane and methylene chloride |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...