Nuclear extract from HeLa cells
Lieferant: BIOMOL RESEARCH LABORATORIES
KI-140-0100
BMOLKI-140-0100EA
0
EUR
BMOLKI-140-0100
Nuclear extract from HeLa cells
Zellen
Nuclear extract from HeLa cells is a rich source of HDAC activity which is used for inhibitor screening or immunoprecipitation of HDAC containing complexes.