Anti-VAMP2 Mouse Monoclonal Antibody

Lieferant: BIOMOL RESEARCH LABORATORIES
VAM-SV014C
BMOLVAM-SV014CEA 0 EUR
BMOLVAM-SV014C
Anti-VAMP2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: VAMP2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR