Reagenzglashalter
Lieferant: BOCHEM
2283
BOCH2283EA 10.9 EUR
BOCH2283
Reagenzglashalter
Zangen
This test tube holder is made from stainless steel (18/10) and features a wooden handle. For tubes with a diameter from 10 to 25 mm.