Universalschere mit Kunststoffgriff
Lieferant: BOCHEM
4251
BOCH4251EA 34.5 EUR
BOCH4251
Universalschere mit Kunststoffgriff
Scheren
Universal scissors with plastic handle (PP + TPE) and stainless steel blades for industrial applications.
  • Adjustable knob: adjustable to fit any hand comfortability
  • Hanging hole: easy to carry
  • Blades made of 4 mm stainless steel (HCR 50-54)
  • Serrated blade for gripping materials
Suitable for cutting aluminium and plastic board, brass plates (non-hardened), leather and cardboard.