Tubes, stainless steel
Lieferant: BOCHEM
500-500 500-700 500-1250 500-1850 500-1000 500-1500 500-300 500-400 500-800
BOCH500-500EA 30.7 EUR
BOCH500-500 BOCH500-700 BOCH500-1250 BOCH500-1850 BOCH500-1000 BOCH500-1500 BOCH500-300 BOCH500-400 BOCH500-800
Tubes, stainless steel
Stativstäbe
Tubes are made of 18/10 stainless steel with grinded surface.
  • Very good chemical resistance
  • Very good temperature resistance