Universal bosshead
Lieferant: BOCHEM
5334
BOCH5334EA 17.2 EUR
BOCH5334
Universal bosshead
Klemmen Stativmuffen
Universal aluminium bosshead with thumb screw (M8).
  • For rods up to 13 mm
  • Angle: 180 + 90°