Double bossheads
Lieferant: BOCHEM
5350 5352 5356 5357 5353 5354
BOCH5350EA 14.6 EUR
BOCH5350 BOCH5352 BOCH5356 BOCH5357 BOCH5353 BOCH5354
Double bossheads
Klemmen Stativmuffen
Double bossheads, either made of stainless steel or aluminium.
  • Angle: 180 + 90°
  • Different screw types available