Double bossheads
Lieferant: BOCHEM
5337 5365
241-3177EA 19.7 EUR
241-3177 BOCH5365
Double bossheads
Klemmen Stativmuffen
Double bossheads, either made of aluminium or malleable cast iron, chromium plated.
  • For rods up to 16,5 mm
  • Angle: 90°
  • With safety screw type 'heart' (M8)
Standards: DIN 12895