Mikroklemmen
Lieferant: BOCHEM
5475
BOCH5475EA 25.3 EUR
BOCH5475
Mikroklemmen
Klemmen Klemmen für den allgemeinen Gebrauch
Micro clamps made of zinc plated steel.
  • Fingers of the clamps are coated with silicone to protect clamped items