Burette stand, DUPLEX
Lieferant: BOCHEM
5620
BOCH5620EA 93 EUR
BOCH5620
Burette stand, DUPLEX
Stative
This burette stand is made of green varnished cast iron.
  • Incl. stainless steel rod (600×10 mm)
  • Incl. two plates with diameter 105 mm (black and white)
  • For use with burette holder (article no. 5600)