Burette and thermometer clamp with shaft, AUTOMATIC
Lieferant: BOCHEM
5624
BOCH5624EA 21.8 EUR
BOCH5624
Burette and thermometer clamp with shaft, AUTOMATIC
Klemmen Klemmen für den allgemeinen Gebrauch
This chromium plated clamp with shaft (length 115 mm) is used for holding burettes or thermometer (Ø 0 - 25 mm).