Double bosshead, Jumbo
Lieferant: BOCHEM
5650
BOCH5650EA 61.5 EUR
BOCH5650
Double bosshead, Jumbo
Klemmen Stativmuffen
Double bossheads made of chrome plated aluminium.
  • For rods up to 20/30 mm
  • Angle: 90°
  • Thread: 10