Sterilisation Boxes for Pipettes
Lieferant: BOCHEM
8980 8981 8982 8983 8984 8985
BOCH8980EA 124 EUR
BOCH8980 BOCH8981 612-1622 BOCH8983 BOCH8984 BOCH8985
Sterilisation Boxes for Pipettes
Boxen Pipetten-Sterilisationsbehälter
Stainless steel.
  • Rotating the lid opens or closes the box
  • Sterilisable up to 205 °C
  • Sturdy
Round sterilising pipette cans with silicone covered bottom and lid.