Overhead stirrer tool, radial stirrer
Lieferant: BOCHEM
9030
BOCH9030EA 98.4 EUR
BOCH9030
Overhead stirrer tool, radial stirrer
Rührer Rührwerkzeuge
Radial stirrer made of stainless steel.
  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low and medium viscous media
  • Creates radial flow
  • Ideal for aeration of liquids