Retort clamp, three pronged
Lieferant: BOCHEM
B08256108
BOCHB08256108EA 141 EUR
BOCHB08256108
Retort clamp, three pronged
Klemmen Klemmen für den allgemeinen Gebrauch
Three pronged retort clamp made of aluminium alloy.
  • Span width is independently adjustable by two regulating screws
  • Jaws are covered with a PVC coating