Retort clamp, four pronged
Lieferant: BOCHEM
B08297100
BOCHB08297100EA 261 EUR
BOCHB08297100
Retort clamp, four pronged
Klemmen Klemmen für den allgemeinen Gebrauch
Stainless steel (18/10)/silicone.
  • Span width is independently adjustable by two regulating screws
  • Four-pronged