Reaktionsgefäße
Lieferant: BOETTGER PAUL
02.013.0100
BOET02.013.0100EA 44.2 EUR
BOET02.013.0100
Reaktionsgefäße
Röhrchen Reaktionsgefäße
Made from selected high-quality and biologically inert polypropylene.
  • High resistance to temperatures and chemicals
  • Autoclavable at 121°C/2,1 bar (cap not closed)
  • Place for markings on label area and flat cover
  • Perfect closure of leak free cap, due to the design of the lid
  • Membrane can be punctured by sampling-needles
  • Graduation
  • Round inside bottom for easy recovery of pellets or fluids
Without caps.