Vials for cell counters
Lieferant: BOETTGER PAUL
02.901.2100
BOET02.901.2100EA 56.6 EUR
BOET02.901.2100
Vials for cell counters
Vials
Optically clear vials with parallel surfaces, free of dust or particles.
  • Covers fitted of optimal density
  • Reliable high quality-level