Sampling pumps, BOLA
Lieferant: Bohlender
A124-16
BOHLA124-16EA 212 EUR
BOHLA124-16
Sampling pumps, BOLA
Pumpen Fass/Transferpumpen
Sampling pump is made up of PP and PTFE.
  • Compact construction and easy handling
  • Quick and low-risk pumping of ultrapure liquids
  • Ergonomic and usable anywhere
  • Universal chemical resistance
A pull on the ball handle produces a slight vacuum in the sampling bottle. Due to this vacuum, the sample is sucked into the sampling bottle.
Glass bottles and plastic bottles with a GL 45 thread can be used as sampling bottles.
Applicable for pumping liquids from sources that are not easily accessible; also, suitable for liquids with a high viscosity like oils.More Product Information