Abdampfschalen, Material:PFA, zylindrisch, porenfrei, stapelbar, Inhalt: 100 ml, Innen Ø 100 mm, Außen Ø 105 mm, Höhe 19,5 mm ,Fl uorkunststoff, 1 * 1 ST
Lieferant: Bohlender
DISH EVAPORATING, 100ML, OUTER DIAMETER 105MM, INNER DIAMETER 100MM, CYLINDRICAL SHAPE, NON-POROUS, STACKABLE, PERFLUOROALKYOXY 1 * 1 items