Distilling heads, BOLA
Lieferant: Bohlender
B289-03
BOHLB289-03EA 320 EUR
BOHLB289-03
Distilling heads, BOLA
Destillationsbauteile Destillieraufsätze
PTFE, white.
  • Temperature resistance: from −20 to +230 °C
  • Universal chemical resistance
Two ground joint cones size NS 29 and one vertical connection with flexible PTFE/Silicone sealing and PPS screw cap to insert and fix thermomometers of a Ø of 7 to 8 mm. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.