Multiple distributors for bottles, BOLA
Lieferant: Bohlender
D651-08
BOHLD651-08EA 129 EUR
BOHLD651-08
Multiple distributors for bottles, BOLA
Verschlüsse Flaschenverschlüsse
Blue screw cap and distributor body both made of PTFE.
  • Very good chemical resistance
  • Temperature resistant from −20 to +200 °C
  • Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with 6 mm external diameter can be inserted through the necks.