Thermoelectric flanging tools, BOLA
Lieferant: Bohlender
F709-50 F709-52 F709-56 F701-12 F709-54 F701-14
BOHLF709-50EA 70.9 EUR
BOHLF709-50 BOHLF709-52 BOHLF709-56 BOHLF701-12 BOHLF709-54 BOHLF701-14
Thermoelectric flanging tools, BOLA
Thermoelectric flanging tools are used for making flanges at the ends of plastic tubing such as PTFE, PFA and FEP tubings.
Lieferumfang: Supplied with basic flanging tool (230 V), exchangeable flanging tip for Int.Ø 0,8 or 1,6 mm and tubing holder for tubing Ext.Ø 1,6 and 3,2 mm.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable