Universal couplings, BOLA
Lieferant: Bohlender
F772-08 F770-08
BOHLF772-08EA 42.3 EUR
BOHLF772-08 BOHLF770-08
Universal couplings, BOLA
Befestigungsmaterial Schrauben
PTFE, white.
  • Temperature resistance: from –200 to +250 °C
  • Universal chemical resistance
  • FDA conform
Coupling for transition from M6 thread to UNF ¼" 28 G thread. The M6 thread is marked by a groove.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable