Ground joint reducing set, BOLA
Lieferant: Bohlender
H981-14
BOHLH981-14EA 90.8 EUR
BOHLH981-14
Ground joint reducing set, BOLA
Adapter Übergangsstücke mit Kegelschliffen
Ground joint reducing set, PTFE, consisting of reducing rings graduated for bridging different cone and socket sizes.
  • Universal chemical resistance
  • Temperature resistance –200 to +250 °C
  • Suitable for vacuum applications