Gas inlet tubes, BOLA
Lieferant: Bohlender
N1502-02 N1502-04 N1502-06
BOHLN1502-02EA 19.7 EUR
BOHLN1502-02 BOHLN1502-04 BOHLN1502-06
Gas inlet tubes, BOLA
Röhrchen Gaseinleitungsrohre
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
For constructing a gas washing equipment. Tube with Int.Ø 5 mm, one side with hose connector dia. 9 mm for connecting tubing, other side with thread M 8×1 for connecting a gas frit or a gas distributor.