Total immersion temperature probes, Lemo®, BOLA
Lieferant: Bohlender
P1790-20
BOHLP1790-20EA 234 EUR
BOHLP1790-20
Total immersion temperature probes, Lemo®, BOLA
Temperaturfühler
Total immersion PTFE probe (class A, PRT 100), with white PTFE-coated cable.
  • Coupling (type Lemo®, socket size 1, 4- pole) and collar ring Ø 5 mm
  • Typical response times of T 50 is 4 to 6 seconds and T 90 is 6 to 8 seconds
Used for measuring temperatures in autoclaves and can be immersed totally into the liquid.
Standards: FDA confirmed.