Tubing cutter, BOLA
Lieferant: Bohlender
S1852-28
BOHLS1852-28EA 68.3 EUR
BOHLS1852-28
Tubing cutter, BOLA
Schneidewerkzeuge
Ideal for cutting plastic and rubber tubing with and without textile reinforcement.
  • Blade is exchangeable
  • Not suitable for steel reinforced tubing