Humidity-temperature probe with USB-interface, BOLA
Lieferant: Bohlender
V1868-08
BOHLV1868-08EA 790 EUR
BOHLV1868-08
Humidity-temperature probe with USB-interface, BOLA
Temperaturfühler
Combined temperature and humidity probe can be directly driven by a PC by means of USB interface in handle.
  • Wide measuring range
  • High resolution
  • Excellent linearity and durability
It is used for monitoring store rooms in food production industry, quality control or HVAC technology, humidity measuring systems for custom projects, microcontroller applications, Windows or Linux.
Used to calculate and indicate the dew point, absolute humidity, vapor pressure, saturated vapor pressure and enthalpy, tabulated measured values and those data are recorded on hard disk, humidity is measured with a long-term stable, capacitive polymer sensor.
Lieferumfang: Includes carry case and windows-software for measured value display and data recording as well as USB plug type A, 1.1 or 2.0 compatible.