Anti-IDE Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-0018R-A647
BOSSBS-0018R-A647EA 893 EUR
BOSSBS-0018R-A647
Anti-IDE Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Plays a role in the cellular breakdown of insulin, IAPP, glucagon, bradykinin, kallidin and other peptides, and thereby plays a role in intercellular peptide signaling. Degrades amyloid formed by APP and IAPP. May play a role in the degradation and clearance of naturally secreted amyloid beta-protein by neurons and microglia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IDE
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR