Anti-IDE Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0018R-A680
BOSSBS-0018R-A680EA 890 EUR
BOSSBS-0018R-A680
Anti-IDE Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Plays a role in the cellular breakdown of insulin, IAPP, glucagon, bradykinin, kallidin and other peptides, and thereby plays a role in intercellular peptide signaling. Degrades amyloid formed by APP and IAPP. May play a role in the degradation and clearance of naturally secreted amyloid beta-protein by neurons and microglia.

Type: Primary
Antigen: Insulin-degrading enzyme
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 491-590/1019
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR